Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159626.1 | NP_001153098.1 | 4 | 390 | ||
NM_024284.2 | NP_077246.2 | 4 | 480 | |||
GenBank mRNA | AK147022.1 | - | 4 | 441 | ||
AK150557.1 | - | 4 | 398 | |||
AK151231.1 | - | 4 | 398 | |||
AK152742.1 | - | 4 | 398 | |||
AK165777.1 | - | 4 | 384 | |||
BC004749.1 | - | 4 | 423 | 3583919 | ||
BC019817.1 | - | 4 | 373 | 5123648 |