Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Stfa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025288.2 | NP_079564.1 | 2 | 171 | ||
GenBank mRNA | AK014349.1 | - | 2 | 174 | ||
AY169236.1 | - | 2 | 154 | |||
BC107224.1 | - | 2 | 165 | 40051891 | ||
BC107225.1 | - | 2 | 167 | 40051892 | ||
M92419.1 | - | 2 | 155 |