Search Thermo Fisher Scientific
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024432.2 | NP_077752.1 | 6 | 649 | ||
GenBank mRNA | AF272895.1 | - | 6 | 664 | ||
AK008942.1 | - | 6 | 658 | |||
AK009117.1 | - | 5 | 431 | |||
AK075705.1 | - | 5 | 837 | |||
AK082510.1 | - | 688 | ||||
AK131187.1 | - | 1 | 1545 | |||
AK148170.1 | - | 617 | ||||
AK161273.1 | - | 6 | 670 | |||
BC016412.1 | - | 6 | 578 | 4215883 | ||
BC049963.1 | - | 6 | 649 | 2655935 |