Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159730.1 | NP_001153202.1 | 4 | 394 | ||
NM_024458.2 | NP_077778.1 | 4 | 382 | |||
GenBank mRNA | AK020798.1 | - | 4 | 383 | ||
BC012507.1 | - | 4 | 368 | 4500900 | ||
BC021920.1 | - | 4 | 383 | 4506350 | ||
BC037176.1 | - | 3 | 2497 | |||
BI738229.1 | - | 4 | 385 | |||
L08075.1 | - | 4 | 401 |