Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025295.4 | NP_079571.1 | 4 | 526 | ||
GenBank mRNA | AK005506.1 | - | 4 | 519 | ||
AK135375.1 | - | 4 | 516 | |||
BC024051.1 | - | 4 | 493 | 5099802 |