Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spaca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024279.2 | NP_077241.1 | 4 | 410 | ||
GenBank mRNA | AK018926.1 | - | 4 | 411 | ||
BC048522.1 | - | 4 | 410 | 6741699 |