Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025387.3 | NP_079663.1 | 4 | 375 | ||
GenBank mRNA | AK003891.1 | - | 4 | 324 | ||
AK008495.1 | - | 3 | 265 | |||
AK008878.1 | - | 4 | 297 | |||
AK010609.1 | - | 4 | 335 | |||
AK012101.1 | - | 4 | 322 | |||
AK019395.1 | - | 4 | 324 | |||
AK050688.1 | - | 4 | 310 | |||
AK089021.1 | - | 3 | 264 | |||
BC025854.1 | - | 4 | 276 | 5250659 | ||
BC055862.1 | - | 4 | 299 | 3492090 | ||
BC069899.1 | - | 3 | 1021 |