Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025336.1 | NP_079612.1 | 8 | 1130 | ||
GenBank mRNA | AK002889.1 | - | 8 | 1202 | ||
AK003519.1 | - | 8 | 1130 | |||
BC032987.1 | - | 8 | 1145 |