Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Akr7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025337.3 | NP_079613.3 | 5 | 769 | ||
GenBank mRNA | AF525358.2 | - | 5 | 671 | ||
AJ271800.1 | - | 5 | 686 | |||
AJ271801.1 | - | 5 | 686 | |||
AK002664.1 | - | 5 | 749 | |||
AK003915.1 | - | 5 | 756 | |||
AK143203.1 | - | 5 | 768 | |||
AK179820.1 | - | 2 | 122 | |||
BC031857.1 | - | 5 | 746 | 5124841 |