Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025388.2 | NP_079664.1 | 7 | 758 | ||
GenBank mRNA | AK003070.1 | - | 6 | 660 | ||
AK003897.1 | - | 7 | 759 | |||
AK011260.1 | - | 6 | 649 | |||
BC021936.1 | - | 7 | 719 | 5124904 | ||
BC055109.1 | - | 2 | 1549 | |||
Y17677.1 | - | 1 | 146 |