Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pithd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025411.4 | NP_079687.3 | 6 | 1289 | ||
GenBank mRNA | AF400670.1 | - | 6 | 1252 | ||
AK004208.1 | - | 6 | 1286 | |||
AK013877.1 | - | 4 | 949 | |||
AK050261.1 | - | 6 | 1262 | |||
BC052695.1 | - | 6 | 1189 | 5715038 |