Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000136.2 | NP_000127.2 | 2 | 342 | ||
NM_001243743.1 | NP_001230672.1 | 2 | 284 | |||
NM_001243744.1 | NP_001230673.1 | 2 | 342 | |||
XM_006717001.2 | XP_006717064.1 | 2 | 324 | |||
XM_006717002.3 | XP_006717065.1 | 2 | 324 | |||
XM_006717004.3 | XP_006717067.1 | 2 | 324 | |||
XM_011518365.2 | XP_011516667.1 | 2 | 317 | |||
XM_011518366.2 | XP_011516668.1 | 2 | 324 | |||
XM_011518367.2 | XP_011516669.1 | 2 | 309 | |||
XM_017014452.1 | XP_016869941.1 | 2 | 324 | |||
XM_017014453.1 | XP_016869942.1 | 2 | 271 | |||
XM_017014454.1 | XP_016869943.1 | 2 | 363 | |||
GenBank mRNA | AK222871.1 | - | 2 | 304 | ||
AK304887.1 | - | 2 | 342 | |||
AK310599.1 | - | 2 | 260 | |||
AK312548.1 | - | 2 | 208 | |||
AU132608.1 | - | 2 | 342 | |||
BC015748.1 | - | 2 | 332 | 4864916 | ||
DB445119.1 | - | 2 | 286 | |||
KU178051.1 | - | 1 | 80 | |||
KU178052.1 | - | 1 | 80 | |||
X66893.1 | - | 2 | 335 | |||
X66894.1 | - | 2 | 253 |