Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam134b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034851.2 | NP_001030023.1 | 9 | 1383 | ||
NM_001277315.1 | NP_001264244.1 | 7 | 1472 | |||
NM_001277316.1 | NP_001264245.1 | 7 | 1055 | |||
NM_001277317.1 | NP_001264246.1 | 7 | 1033 | |||
NM_001277318.1 | NP_001264247.1 | 7 | 1005 | |||
NM_025459.3 | NP_079735.2 | 8 | 1173 | |||
GenBank mRNA | AK007506.1 | - | 7 | 1034 | ||
AK015182.1 | - | 8 | 995 | |||
AK015887.1 | - | 1 | 352 | |||
AK146972.1 | - | 7 | 1056 | |||
AK167327.1 | - | 7 | 1473 | |||
BC019494.1 | - | 8 | 1112 | 4460992 | ||
BC054415.1 | - | 7 | 992 | 3487542 |