Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025532.3 | NP_079808.1 | 2 | 207 | ||
GenBank mRNA | AF525155.1 | - | 2 | 158 | ||
AK005411.1 | - | 2 | 208 | |||
AK009863.1 | - | 2 | 208 | |||
BC100329.1 | - | 2 | 197 | 30787440 | ||
BC111112.1 | - | 2 | 203 | 40050224 | ||
BC138199.1 | - | 2 | 200 | 8861218 |