Gene Symbol: Nipsnap3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025623.2 | NP_079899.1 | 2 | 161 | ||
GenBank mRNA | AK004458.1 | - | 2 | 160 | ||
AK008786.1 | - | 2 | 157 | |||
AK012480.1 | - | 2 | 161 | |||
BC019505.1 | - | 2 | 146 | 4164310 |