Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025531.2 | NP_079807.1 | 3 | 352 | ||
GenBank mRNA | AK003043.1 | - | 3 | 326 | ||
AK008542.1 | - | 3 | 312 | |||
AK013197.1 | - | 3 | 319 | |||
AK150668.1 | - | 3 | 328 | |||
BC004775.1 | - | 2 | 150 | |||
BC026968.1 | - | 3 | 352 | 4235395 |