Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025539.2 | NP_079815.2 | 3 | 528 | ||
GenBank mRNA | AK009933.1 | - | 3 | 526 | ||
AK018771.1 | - | 3 | 526 | |||
AK132532.1 | - | 529 | ||||
BC025153.1 | - | 2 | 504 | 4236805 |