Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: APOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000042.2 | NP_000033.2 | 2 | 242 | ||
GenBank mRNA | AK298416.1 | - | 2 | 214 | ||
AK313838.1 | - | 2 | 198 | |||
BC020703.1 | - | 2 | 200 | 4767966 | ||
BC026283.1 | - | 2 | 214 | 4734357 | ||
BG566874.1 | - | 2 | 242 | |||
HM212425.1 | - | 1 | 126 | |||
M62839.1 | - | 2 | 222 | |||
S80305.1 | - | 2 | 192 | |||
X53595.1 | - | 2 | 216 | |||
X57847.1 | - | 2 | 216 | |||
X58100.1 | - | 2 | 204 |