Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025550.2 | NP_079826.2 | 3 | 466 | ||
GenBank mRNA | AJ252147.1 | - | 3 | 502 | ||
AK002343.1 | - | 3 | 500 | |||
AK010287.1 | - | 3 | 499 | |||
AK217031.1 | - | 1 | 37 | |||
BC006869.1 | - | 3 | 466 | 2646325 |