Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Srsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025573.3 | NP_079849.1 | 4 | 825 | ||
GenBank mRNA | AK011621.1 | - | 4 | 785 | ||
AK020084.1 | - | 4 | 597 | |||
AK159190.1 | - | 4 | 823 | |||
BC012217.1 | - | 4 | 825 | 3483731 | ||
CV558546.1 | - | 4 | 574 |