Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 2810428I15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025577.2 | NP_079853.1 | 4, 5 | 452 | ||
GenBank mRNA | AK010635.1 | - | 4, 5 | 449 | ||
AK013181.1 | - | 4, 5 | 453 | |||
AK160511.1 | - | 4, 5 | 396 | |||
AK180760.1 | - | 2, 3 | 129 | |||
BC028495.1 | - | 4, 5 | 458 | 3370477 |