Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025611.5 | NP_079887.3 | 25 | 5058 | ||
GenBank mRNA | AK008655.1 | - | 2 | 246 | ||
AK010907.1 | - | 4 | 583 | |||
AK147156.1 | - | 25 | 4816 | |||
AK170808.1 | - | 25 | 4831 | |||
AK172882.1 | - | 24 | 4131 | |||
BC019645.1 | - | 15 | 2346 | |||
BC024345.1 | - | 4 | 850 | |||
BC026946.1 | - | 7 | 1166 | |||
BC059865.1 | - | 21 | 3571 | |||
BC065096.1 | - | 24 | 4545 | |||
BC100544.1 | - | 4 | 694 | |||
BC138998.1 | - | 25 | 4996 | 8862020 | ||
BC139000.1 | - | 25 | 4996 | 8862022 |