Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025745.3 | NP_080021.3 | 2, 3 | 331 | ||
GenBank mRNA | AK002471.1 | - | 2, 3 | 322 | ||
AK077120.1 | - | 2, 3 | 324 | |||
AK133874.1 | - | 2, 3 | 332 | |||
BC018468.1 | - | 2, 3 | 341 | 3591901 |