Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025705.3 | NP_079981.2 | 2 | 361 | ||
GenBank mRNA | AK014521.1 | - | 2 | 173 | ||
AK016485.1 | - | 2 | 361 | |||
AK157211.1 | - | 2 | 348 |