Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162538.1 | NP_001156010.1 | 3 | 483 | ||
NM_025714.4 | NP_079990.2 | 3 | 349 | |||
GenBank mRNA | AK014630.1 | - | 3 | 351 | ||
AK016726.1 | - | 3 | 465 | |||
AK031564.1 | - | 3 | 485 | |||
AK048931.1 | - | 3 | 465 | |||
AK085115.1 | - | 3 | 473 | |||
BB621104.1 | - | 3 | 485 |