Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Emc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025736.2 | NP_080012.1 | 4 | 345 | ||
GenBank mRNA | AF332065.1 | - | 4 | 273 | ||
AF332066.1 | - | 4 | 273 | |||
AK014989.1 | - | 4 | 346 | |||
AK015968.1 | - | 4 | 311 | |||
AK172887.1 | - | 4 | 290 | |||
BC004716.1 | - | 4 | 303 | 3582196 |