Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Henmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078646.1 | NP_001072114.1 | 8 | 1617 | ||
NM_025723.2 | NP_079999.2 | 8 | 1932 | |||
GenBank mRNA | AK038994.1 | - | 8 | 1618 | ||
AK085587.1 | - | 8 | 1931 | |||
BC099408.1 | - | 7 | 1598 | 30935249 |