Search Thermo Fisher Scientific
Gene Symbol: Emc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025736.2 | NP_080012.1 | 11 | 893 | ||
GenBank mRNA | AF332065.1 | - | 11 | 821 | ||
AF332066.1 | - | 11 | 822 | |||
AK014989.1 | - | 11 | 894 | |||
AK015968.1 | - | 11 | 859 | |||
AK172887.1 | - | 11 | 838 | |||
BC004716.1 | - | 11 | 851 | 3582196 |