Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025850.2 | NP_080126.1 | 3 | 281 | ||
GenBank mRNA | AK005694.1 | - | 3 | 291 | ||
AK017071.1 | - | 3 | 305 | |||
AY819025.1 | - | 3 | 311 | |||
BC046278.1 | - | 3 | 281 | 6310911 |