Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025794.2 | NP_080070.2 | 3 | 509 | ||
GenBank mRNA | AK002483.1 | - | 3 | 462 | ||
AK028686.1 | - | 3 | 461 | |||
AK037969.1 | - | 3 | 426 | |||
AK075673.1 | - | 3 | 460 | |||
AK085493.1 | - | 3 | 478 | |||
AK141684.1 | - | 3 | 463 | |||
AK152624.1 | - | 3 | 478 | |||
AK169686.1 | - | 3 | 414 | |||
BC012522.1 | - | 3 | 458 | 2648103 | ||
BC057670.1 | - | 3 | 476 | 2810976 |