Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033573.1 | NP_001028745.1 | 2 | 726 | ||
NM_025883.3 | NP_080159.3 | 2 | 610 | |||
GenBank mRNA | AK010296.1 | - | 2 | 605 | ||
AK011373.1 | - | 2 | 577 | |||
AK035172.1 | - | 689 | ||||
AK038177.1 | - | 589 | ||||
AK050388.1 | - | 2 | 725 | |||
AK077693.1 | - | 2 | 564 | |||
AK077838.1 | - | 575 | ||||
AK139707.1 | - | 2 | 567 | |||
AK144128.1 | - | 2 | 582 | |||
AK154741.1 | - | 2 | 588 | |||
AK157494.1 | - | 2 | 592 | |||
AK159304.1 | - | 2 | 611 | |||
AK168497.1 | - | 2 | 605 | |||
AK179827.1 | - | 2 | 389 | |||
AK181252.1 | - | 2 | 389 | |||
AK185086.1 | - | 2 | 398 | |||
AK208744.1 | - | 1 | 285 | |||
AK210763.1 | - | 1 | 294 | |||
BC033317.1 | - | 2 | 557 | 4459242 |