Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012312.1 | - | 1 | 119 | ||
AK016651.1 | - | 1 | 262 | |||
AK018787.1 | - | 1 | 129 | |||
AK038578.1 | - | 1 | 83 | |||
AK044291.1 | - | 1 | 232 | |||
AK048044.1 | - | 1 | 232 | |||
AK076231.1 | - | 1 | 246 | |||
AK078139.1 | - | 1 | 230 | |||
AK078468.1 | - | 1 | 246 | |||
AK078872.1 | - | 1 | 163 | |||
AK079556.1 | - | 1 | 83 | |||
AK080238.1 | - | 1 | 128 | |||
AK161465.1 | - | 1 | 142 | |||
AK165398.1 | - | 1 | 248 | |||
BC031761.1 | - | 1 | 112 | 4235549 |