Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Magt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190409.1 | NP_001177338.1 | 1, 2 | 243 | ||
NM_025952.4 | NP_080228.4 | 1, 2 | 243 | |||
GenBank mRNA | AK010320.1 | - | 1, 2 | 179 | ||
AK012185.1 | - | 1, 2 | 120 | |||
AK013243.1 | - | 1, 2 | 124 | |||
AK018623.1 | - | 1, 2 | 146 | |||
AK136622.1 | - | 1, 2 | 125 | |||
AK144696.1 | - | 1, 2 | 122 | |||
BC003881.1 | - | 1, 2 | 130 | 3496572 | ||
DQ000005.1 | - | 1, 2 | 243 |