Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lce1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025984.2 | NP_080260.1 | 2 | 622 | ||
GenBank mRNA | AK008643.2 | - | 2 | 623 | ||
AK009102.1 | - | 2 | 624 | |||
BC108406.1 | - | 2 | 598 |