Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290500.1 | NP_001277429.1 | 5 | 683 | ||
NM_001290502.1 | NP_001277431.1 | 5 | 921 | |||
NM_025978.4 | NP_080254.2 | 5 | 683 | |||
NM_027619.4 | NP_081895.2 | 5 | 683 | |||
GenBank mRNA | AK012246.1 | - | 5 | 645 | ||
AK015308.1 | - | 5 | 643 | |||
AK016619.1 | - | 5 | 603 | |||
AK017763.1 | - | 3 | 281 | |||
AK031605.1 | - | 5 | 914 | |||
AK035570.1 | - | 5 | 653 | |||
AK041095.1 | - | 5 | 657 | |||
AK044266.1 | - | 5 | 658 | |||
AK134539.1 | - | 5 | 672 | |||
AK173315.1 | - | 5 | 653 | |||
BC021448.1 | - | 5 | 691 | |||
BC024847.1 | - | 5 | 648 | 5355806 | ||
BC138212.1 | - | 5 | 645 | 8861231 | ||
BC145944.1 | - | 5 | 645 | 40131247 |