Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zcchc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035509.1 | NP_001030586.1 | 4 | 1391 | ||
NM_001035510.1 | NP_001030587.1 | 2 | 1862 | |||
NM_025893.2 | NP_080169.2 | 4 | 1339 | |||
GenBank mRNA | AK005317.1 | - | 4 | 1317 | ||
AK075607.1 | - | 1 | 943 | |||
AK134856.1 | - | 1720 | ||||
AY650116.1 | - | 1 | 821 | |||
BC017627.1 | - | 4 | 1252 | 4505033 |