Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025903.2 | NP_080179.1 | 12 | 1811 | ||
GenBank mRNA | AK002702.1 | - | 6 | 1036 | ||
AK007687.1 | - | 12 | 1812 | |||
AK149496.1 | - | 12 | 1792 | |||
AK156625.1 | - | 11 | 1887 | |||
AK165967.1 | - | 12 | 1804 | |||
AK166130.1 | - | 12 | 1879 | |||
AK166258.1 | - | 12 | 1769 | |||
BC023486.1 | - | 12 | 1758 | |||
BC037434.1 | - | 12 | 1767 | 2651172 |