Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025907.3 | NP_080183.1 | 4, 5 | 758 | ||
GenBank mRNA | AK005448.1 | - | 4, 5 | 813 | ||
AK006715.1 | - | 3, 4 | 665 | |||
AK078185.1 | - | 4, 5 | 812 | |||
AK082949.1 | - | 4, 5 | 814 | |||
AK133735.1 | - | 4, 5 | 790 | |||
AK189650.1 | - | 18 | ||||
BC030449.1 | - | 4, 5 | 759 | 5400506 |