Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000042.2 | NP_000033.2 | 7 | 976 | ||
GenBank mRNA | AK298416.1 | - | 7 | 735 | ||
AK313838.1 | - | 7 | 932 | |||
BC020703.1 | - | 7 | 934 | 4767966 | ||
BC026283.1 | - | 7 | 948 | 4734357 | ||
BM005736.1 | - | 2 | 331 | |||
HM212425.1 | - | 6 | 860 | |||
M62839.1 | - | 7 | 956 | |||
S80305.1 | - | 7 | 926 | |||
X53595.1 | - | 7 | 950 | |||
X57847.1 | - | 7 | 950 | |||
X58100.1 | - | 7 | 938 |