Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdnep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026017.2 | NP_080293.1 | 3 | 498 | ||
GenBank mRNA | AK012063.1 | - | 3 | 480 | ||
AK076150.1 | - | 3 | 512 | |||
AK164602.1 | - | 3 | 440 | |||
BC018265.1 | - | 3 | 498 | 4486464 |