Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mrpl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026065.3 | NP_080341.1 | 4 | 283 | ||
GenBank mRNA | AK002530.1 | - | 4 | 268 | ||
AK003427.2 | - | 4 | 284 | |||
AK012227.1 | - | 4 | 268 | |||
AK013693.1 | - | 4 | 280 | |||
AK154032.1 | - | 4 | 279 | |||
BC024337.1 | - | 4 | 264 | 4984291 |