Gene Symbol: Iqcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026090.2 | NP_080366.2 | 2 | 173 | ||
GenBank mRNA | AK006688.1 | - | 2 | 172 | ||
AK007079.1 | - | 2 | 203 | |||
AK018930.1 | - | 2 | 203 | |||
BC048652.1 | - | 2 | 178 | 6772797 | ||
BC061014.1 | - | 2 | 173 | 6770556 | ||
DQ153246.1 | - | 2 | 202 |