Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713314.3 | XP_006713377.1 | 5 | 1180 | ||
XM_011534041.2 | XP_011532343.1 | 5 | 1017 | |||
XM_017007088.1 | XP_016862577.1 | 6 | 1293 | |||
GenBank mRNA | AK294301.1 | - | 6 | 1032 | ||
AK297033.1 | - | 4 | 892 | |||
AK301838.1 | - | 4 | 1092 | |||
AK313252.1 | - | 4 | 883 | |||
BC012099.1 | - | 4 | 826 | 4559618 | ||
U03274.1 | - | 4 | 829 |