Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026014.3 | NP_080290.3 | 4 | 752 | ||
GenBank mRNA | AB086655.1 | - | 4 | 731 | ||
AF477481.1 | - | 4 | 752 | |||
AK012041.1 | - | 4 | 729 | |||
AK017609.1 | - | 4 | 726 | |||
AK028287.1 | - | 4 | 724 | |||
AK049163.1 | - | 4 | 724 | |||
AK076527.1 | - | 4 | 742 | |||
AK083015.1 | - | 4 | 727 | |||
AK194030.1 | - | 2 | 328 | |||
AK198035.1 | - | 1 | 81 | |||
AK204196.1 | - | 1 | 81 | |||
BC013842.1 | - | 4 | 707 | |||
BC048076.1 | - | 4 | 752 | 6307476 |