Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252374.1 | NP_001239303.1 | 7, 8 | 505 | ||
NM_026004.3 | NP_080280.3 | 6, 7 | 662 | |||
GenBank mRNA | AK011894.1 | - | 6, 7 | 560 | ||
AK160139.1 | - | 7, 8 | 505 | |||
BC038029.1 | - | 7, 8 | 498 | 4018956 | ||
CB571852.1 | - | 7, 8 | 510 |