Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026053.3 | NP_080329.1 | 3 | 838 | ||
GenBank mRNA | AK011454.1 | - | 3 | 819 | ||
AK013396.1 | - | 3 | 840 | |||
AK020147.1 | - | 3 | 862 | |||
AK157745.1 | - | 3 | 851 | |||
AK167680.1 | - | 3 | 831 | |||
AK167920.1 | - | 3 | 877 | |||
BC025157.1 | - | 3 | 821 | 3670793 |