Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026147.5 | NP_080423.1 | 2, 3 | 224 | ||
GenBank mRNA | AK002405.1 | - | 2, 3 | 225 | ||
AK014594.1 | - | 224 | ||||
AK136398.1 | - | 2, 3 | 227 | |||
AK145656.1 | - | 2, 3 | 225 | |||
AK150867.1 | - | 2, 3 | 224 | |||
AK150972.1 | - | 2, 3 | 224 | |||
BC011323.1 | - | 2, 3 | 211 | 3257215 | ||
BC090389.1 | - | 2, 3 | 215 | 3482252 |