Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026123.3 | NP_080399.1 | 2 | 519 | ||
GenBank mRNA | AK003273.1 | - | 2 | 294 | ||
AK004433.1 | - | 2 | 283 | |||
AK132757.1 | - | 2 | 520 | |||
AK135885.1 | - | 2 | 511 | |||
AK217920.1 | - | 2 | 242 | |||
AY017214.1 | - | 2 | 504 | |||
BC019484.1 | - | 2 | 486 | 4039903 |