Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026141.3 | NP_080417.2 | 3, 4 | 252 | ||
GenBank mRNA | AK011443.1 | - | 3, 4 | 224 | ||
AK012371.1 | - | 3, 4 | 229 | |||
AK013377.1 | - | 3, 4 | 225 | |||
AK014406.2 | - | 225 | ||||
AK014453.1 | - | 3, 4 | 240 | |||
AK017370.1 | - | 3, 4 | 252 | |||
AK167168.1 | - | 3, 4 | 217 | |||
AK167258.1 | - | 3, 4 | 224 | |||
BC079912.1 | - | 3, 4 | 214 | 30661438 | ||
BY032114.1 | - | 3, 4 | 255 |