Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026141.3 | NP_080417.2 | 4 | 319 | ||
GenBank mRNA | AK011443.1 | - | 4 | 291 | ||
AK012371.1 | - | 4 | 296 | |||
AK013377.1 | - | 4 | 292 | |||
AK014406.2 | - | 292 | ||||
AK014453.1 | - | 4 | 307 | |||
AK017370.1 | - | 4 | 319 | |||
AK167168.1 | - | 4 | 284 | |||
AK167258.1 | - | 4 | 291 | |||
BC079912.1 | - | 4 | 281 | 30661438 | ||
BY032114.1 | - | 4 | 323 |